Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGLUT1 Antibody (S28-9), Novus Biologicals™

Mouse Monoclonal Antibody
$562.50
Specifications
| Antigen | VGLUT1 |
|---|---|
| Clone | S28-9 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
Description
VGLUT1 Monoclonal specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| VGLUT1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| SLC17A7 | |
| IgG1 | |
| Protein G purified |
| S28-9 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 57030 | |
| Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1 | |
| Primary | |
| Detects 52kDa. No cross-reactivity against VGlut2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title