Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGluT3 Antibody, Novus Biologicals™
SDP

Catalog No. NB177048 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177048 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177048 Supplier Novus Biologicals Supplier No. NBP313361
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VGluT3 Polyclonal antibody specifically detects VGluT3 in Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen VGluT3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of mouse VGluT3. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline ABC Transporters, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 246213
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.