Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ VH 101, thiol

Click to view available options
Quantity:
50 mg
Description
VH 101, thiol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with terminal thiol ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
Chemical Name or Material | (2S,4R)-1-[(2R)-2-[[(1-Fluorocyclopropyl)carbonyl]amino]-3-methyl-3-sulfanyl-butanoyl]-4-hydroxy-N-[[4-(4-methylthiazol-5-yl)phenyl]methyl]pyrrolidine-2-carboxamide |
CAS | 2358775-81-0 |
Quantity | 50 mg |
Target | Degrader Building Blocks |
Molecular Formula | C25H31FN4O4S2 |
Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction