Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHR/DUSP3 Antibody (9E6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB161778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161778 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB161778 Supplier Novus Biologicals Supplier No. NBP259460UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VHR/DUSP3 Monoclonal antibody specifically detects VHR/DUSP3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen VHR/DUSP3
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 9000000
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias dual specificity phosphatase 3, dual specificity protein phosphatase 3, Dual specificity protein phosphatase VHR, EC 3.1.3.16, EC 3.1.3.48, Vaccinia H1-related phosphatase, vaccinia virus phosphatase VH1-related, VHRserine/threonine specific protein phosphatase
Host Species Mouse
Immunogen Recombinant human VHR/DUSP3 (1-185aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 1845
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.