Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHR/DUSP3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB056172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056172 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056172 Supplier Novus Biologicals Supplier No. NBP298873AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VHR/DUSP3 Polyclonal specifically detects VHR/DUSP3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen VHR/DUSP3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias dual specificity phosphatase 3, dual specificity protein phosphatase 3, Dual specificity protein phosphatase VHR, EC 3.1.3.16, EC 3.1.3.48, Vaccinia H1-related phosphatase, vaccinia virus phosphatase VH1-related, VHRserine/threonine specific protein phosphatase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant VHR/DUSP3 (P51452; Met1-Pro185).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 1845
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.