Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIAAT/SLC32A1/VGAT Antibody (D9), Novus Biologicals™
SDP

Catalog No. NB010673 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010673 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010673 Supplier Novus Biologicals Supplier No. NBP279823

Mouse Monoclonal Antibody

VIAAT/SLC32A1/VGAT Monoclonal specifically detects VIAAT/SLC32A1/VGAT in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen VIAAT/SLC32A1/VGAT
Applications Western Blot, ELISA
Classification Monoclonal
Clone D9
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT
Gene Symbols SLC32A1
Host Species Mouse
Immunogen Synthetic peptide from the N-terminal region of Mouse VGAT protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.