Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIAAT/SLC32A1/VGAT Antibody (HL1616) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294874 Supplier Novus Biologicals Supplier No. NBP325743
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VIAAT/SLC32A1/VGAT Monoclonal antibody specifically detects VIAAT/SLC32A1/VGAT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen VIAAT/SLC32A1/VGAT
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HL1616
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VIAAT/SLC32A1/VGAT. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 140679
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.