Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIAAT/SLC32A1/VGAT Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP234117
Description
VIAAT/SLC32A1/VGAT Polyclonal specifically detects VIAAT/SLC32A1/VGAT in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VIAAT/SLC32A1/VGAT | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q9H598 | |
SLC32A1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: ATSVSNKSQAKMSGMFARMGFQAATDEEAVGFAHCDDLDFEHRQGLQMDILKAEGEPCGDEGAEAPVEGDIHYQRGSGAPLPPSGSKDQVG | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT | |
Rabbit | |
Affinity Purified | |
RUO | |
140679 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction