Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Victoria Blue B, pure

Catalog No. p-2374489
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 2580-56-5 | C33H32ClN3 | 506.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2580-56-5
Molecular Formula C33H32ClN3
Molecular Weight (g/mol) 506.09
MDL Number MFCD00011878
InChI Key LLWJPGAKXJBKKA-UHFFFAOYSA-N
Synonym Basic Blue 26, C.I. 44045
PubChem CID 17407
ChEBI CHEBI:87651
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]naphthalen-1-ylidene]-phenylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[NH+]C3=CC=CC=C3)C4=CC=CC=C24)C5=CC=C(C=C5)N(C)C.[Cl-]

Specifications

Melting Point 206.0°C
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
Infrared Spectrum Authentic
Lambda Max 599 to 605nm (in ethanol)
Formula Weight 506.09
Loss on Drying 5% max. (60°C)
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Brown
Quantity 25 g
Physical Form Glistening Crystalline Powder
Chemical Name or Material Victoria blue B
Show More Show Less
EINECSNumber 219-943-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.