Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vidaro™ B-Fore™ ESD Hoods
Shop All Vidaro Corporation Products
Click to view available options
Color:
Light Blue
Navy
White
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
Requires
Two female snaps on each cheek allow the attachment of a facial (not included).
- Super-dense weave of 100% microdenier polyester fibers minimizes particulate contamination
- Microengineered carbon fiber grid dissipates electrostatic charges
- Open-faced hood fits snugly under the chin, leaving the entire face exposed
- Full-bib apron is long, to fit securely into a coverall or frock without pulling out.
- Front closure has two stainless-steel snaps, and there are adjustment snaps at the back of the neck
Specifications
Specifications
Product Type | Open Face |
Size | 2X-Large |
Material | Polyester, Carbon |
Color | Navy |
Closure Type | Snap Front |
Disposable | Reusable |
Model | H-4358 |
Quantity | 1 Ea. |
Includes | Two snaps on each cheek, Stainless Steel front-snap closure, snap at back of neck for adjustment |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction