Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ 0.6 in. Drain Cleaning Brush

Catalog No. p-7161957
Click to view available options
Color:
Black
Blue
Red

Vikan™ 0.6 in. Drain Cleaning Brush features stiff bristles mounted on a flexible stainless steel rod.

  • Used to clean small spaces between machine parts and drainage holes in sinks
  • The short handle allows the operator to exert pressure to get drains clean
  • The stiff bristles back the effort up
  • Color-coded

Specifications

Product Type Cleaning Brush
Bristle Material Polyester
Bristle Type Stiff
Color-coded Yes
Handle Material Polypropylene
Sterility Non-sterile
Certifications/Compliance FDA Compliant
Color Red
Diameter (English) Bristle 0.6 in.
Length (English) 12.2 in.
Quantity 1 Ea.
Size Small
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.