Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vikan™ Medium Dish Brush with Scraper
Shop All Remco Products
Click to view available options
Color:
Blue
Green
Orange
Pink
Red
White
Yellow
Description
- Angled bristles that reach into corners
- Inlcudes a small scraper at the tip to loosen stubborn dirt
- The raised angle of the grip promotes quick and effective cleaning
- Color-coded
- Ideal for cleaning cups, bowls, and machinery
Specifications
Specifications
| Product Type | Dish Brush |
| Bristle Material | Polyester |
| Color-coded | Yes |
| Handle Material | Polypropylene |
| Sterility | Non-sterile |
| Certifications/Compliance | FDA Compliant |
| Color | Red |
| Length (English) | 10.8 in. |
| Quantity | 1 Ea. |
| Size | Medium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction