Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Vildagliptin anti-diabetic agent
Shop All Novus Biologicals Products

Click to view available options
Quantity:
20 mg
100 mg
Description
PubChem ID: 11077541
Application Notes: May be dissolved in DMSO (45 mg/mL); or DMF (20 mg/mL); or ethanol (20 mg/mL).
Specifications
Specifications
CAS | 274901-16-5 |
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Molecular Formula | C17H25N3O2 |
Purity | >98% (HPLC); NMR (conforms) |
Quantity | 20 mg |
Source | Synthetic |
Target | Vildagliptin anti-diabetic agent |
Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction