Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villin 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132841 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132841 Supplier Novus Biologicals Supplier No. NBP132841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Villin 1 Polyclonal antibody specifically detects Villin 1 in Human, Mouse, Rat, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen Villin 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen -Assay dependent
Formulation PBS, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Alias D2S1471, VIL, villin 1, villin-1
Gene Symbols VIL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Villin. The exact sequence is proprietary.
Molecular Weight of Antigen 93 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7429
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse, Rat, Canine, Feline
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.