Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villin 1 Mouse anti-Human, Clone: VIL1/1314- Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254516200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254516200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB254516200 Supplier Novus Biologicals Supplier No. NBP254516200UG

Mouse Monoclonal Antibody

Specifications

Antigen Villin 1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone VIL1/1314
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias D2S1471, VIL, villin 1, villin-1
Gene Symbols VIL1
Host Species Mouse
Immunogen Recombinant human Villin fragment of 133 amino acid residues (aa179-311) (exact sequence is proprietary)
Molecular Weight of Antigen 93 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7429
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.