Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Antibody (211), Alexa Fluor™ 594
SDP

Catalog No. NB087753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087753 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087753 Supplier Novus Biologicals Supplier No. NBP289211AF594

Rabbit Monoclonal Antibody

Vimentin Monoclonal specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Vimentin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 211
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias FLJ36605, vimentin
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Vimentin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.