Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Antibody (280618), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB366399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366399 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366399 Supplier Novus Biologicals Supplier No. FAB2105E
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen Vimentin
Applications Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 280618
Conjugate DyLight 405
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Host Species Rat
Immunogen E. coli-derived recombinant human Vimentin, Ser2-Glu466, Accession # P08670
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.