Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (2A52), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP192688
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
Vimentin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Knockout Validated | |
P08670 | |
VIM | |
Full length recombinant human vimentin protein expressed in and purified from E. coli. | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Cow and Porcine. Based upon sequence similarity, this antibody is predicted to react with Chimpanzee (99%), Monkey (99%) and Hamster (97%). Sequence homology of Human Vimentin to Chicken and Xenopus is 86% and 78% respectively. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
2A52 | |
Unconjugated | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
FLJ36605, vimentin | |
Mouse | |
50 kDa | |
0.1 mL | |
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
7431 | |
Human, Rat, Mouse (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction