Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Antibody (2A52), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$243.50 - $499.50
Specifications
Antigen | Vimentin |
---|---|
Clone | 2A52 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP192688SS
![]() |
Novus Biologicals
NBP192688SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP192688
![]() |
Novus Biologicals
NBP192688 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
Vimentin | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
Mouse | |
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
50% PBS, 50% glycerol with 5mM Sodium Azide | |
FLJ36605, vimentin | |
VIM | |
IgG1 | |
Protein G purified | |
Human, Mouse, Rat, Cow and Porcine. Based upon sequence similarity, this antibody is predicted to react with Chimpanzee (99%), Monkey (99%) and Hamster (97%). Sequence homology of Human Vimentin to Chicken and Xenopus is 86% and 78% respectively. |
2A52 | |
Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/Immunofluorescence 1:5000, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Mouse (Negative) | |
P08670 | |
7431 | |
Full length recombinant human vimentin protein expressed in and purified from E. coli. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title