Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Antibody, Novus Biologicals™
SDP

Catalog No. NB414069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
0.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB414069 0.5 mL
NB300223SS 0.025 mL
NB300223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB414069 Supplier Novus Biologicals Supplier No. NB3002230.5ML
Add to Cart
Add to Cart

Chicken Polyclonal Antibody has been used in 44 publications

Vimentin Polyclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Vimentin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:20000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry-Paraffin 1:150, Immunohistochemistry-Frozen
Formulation Supplied as an aliquot of IgY preparation with 5mM Sodium Azide
Gene Accession No. P08670
Gene Alias FLJ36605, vimentin
Gene Symbols VIM
Host Species Chicken
Immunogen Full length recombinant human Vimentin expressed in and purified from E. coli. [Swiss-Prot# P08670]
Molecular Weight of Antigen 50 kDa
Purification Method IgY purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human, Mouse, Rat, Pig, Bovine, Chicken, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.