Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Antibody (RV203), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NBP97671549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP97671549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP97671549 Supplier Novus Biologicals Supplier No. NBP197671JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Goat, Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Vimentin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RV203
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ36605, vimentin
Gene Symbols VIM
Host Species Mouse
Immunogen Clone RV203 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal vimentin extract of calf lens.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Test Specificity RV203 reacts exclusively with vimentin, which is expressed in mesenchymal cells and mesenchymal derived tumors e.g. lymphoma, sarcoma and melanoma.
Target Species Canine, Goat, Hamster, Human, Mouse, Pig, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.