Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Antibody (VIM/1937R), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB389246 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389246 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389246 Supplier Novus Biologicals Supplier No. NBP308936AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Rat, Porcine, Bovine, Canine, Chicken, Equine, Feline, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Vimentin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VIM/1937R
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Host Species Rabbit
Immunogen Recombinant full-length human Vimentin protein (Uniprot: P08670)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human, Rat, Pig, Bovine, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.