Learn More
Description
Specifications
Specifications
| Antigen | Vimentin |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Clone | VM452 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated |
| Formulation | 50mM Sodium Borate |
| Gene Alias | epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin |
| Host Species | Mouse |
| Immunogen | Recombinant human Vimentin protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
