| Antigen |
Vimentin |
| Applications |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification |
Monoclonal |
| Clone |
SPM576 |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
| Formulation |
PBS with 0.05% BSA. with 0.05% Sodium Azide |
| Gene Accession No. |
P08670 |
| Gene Alias |
FLJ36605, vimentin |
| Gene Symbols |
VIM |
| Host Species |
Mouse |
| Immunogen |
Human vimentin recombinant protein |
| Purification Method |
Protein G purified |
| Quantity |
0.1 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
7431 |
| Test Specificity |
This MAb reacts with a 58kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFPs) such as desmin, keratin, neurofilament, and glial fibrillary acid protein.Anti-vimentin alone is of limited value as a diagnostic tool; however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain vimentin, e.g. hepatoma and seminoma. Anti-vimentin is also useful as a tissue process control reagent. |
| Target Species |
Human, Pig, Bovine, Canine, Chicken, Feline, Goat, Mouse (Negative), Rat (Negative) |
| Content And Storage |
Store at 4C. |
| Form |
Purified |
| Isotype |
IgG1 |
|
Show More
Show Less
|
|