Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin, p Ser71 Antibody (TM71), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP1975150.025MG
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Vimentin (p Ser71) | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with No Preservative | |
VIM | |
Synthetic peptide corresponding to aa 66-77 of human Vimentin phosphorylated at Ser71. | |
0.025 mg | |
Phospho Specific, Stem Cells | |
7431 | |
Rat, Mouse, Human | |
Purified |
Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
TM71 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 ug/ml | |
FLJ36605, vimentin | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Recognizes Human, Mouse and Rat Vimentin. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction