Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin, p Ser71 Antibody (TM71), Novus Biologicals™

Rat Monoclonal Antibody
$243.50 - $572.50
Specifications
Antigen | Vimentin (p Ser71) |
---|---|
Clone | TM71 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 ug/ml |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Vimentin (p Ser71) | |
Western Blot 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Rat, Mouse, Human | |
7431 | |
Synthetic peptide corresponding to aa 66-77 of human Vimentin phosphorylated at Ser71. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
TM71 | |
Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rat | |
Phospho Specific, Stem Cells | |
FLJ36605, vimentin | |
VIM | |
IgG2a | |
Protein G purified | |
Recognizes Human, Mouse and Rat Vimentin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title