Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinblastine Sulfate, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J63598MFPM
Click to view available options
Quantity:
50 mg

Anticancer agent; microtubule disrupter; Induces apoptosis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-67-9
Molecular Formula C46H60N4O13S
Synonym Vincaleukblastine sulfate

Specifications

Quantity 50 mg
Content And Storage 2°C to 8°C
For Use With (Application) Anticancer agent; microtubule disrupter; Induces apoptosis
Melting Point 267°C
Solubility Soluble in water or DMSO
Product Type Signal Transduction Reagent
Form Powder
Assay 98%

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.