Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396428 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396428 Supplier Novus Biologicals Supplier No. NBP2412740.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vinculin Polyclonal antibody specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Immunocytochemistry

Specifications

Antigen Vinculin
Applications Western Blot, ELISA, Immunofluorescence, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 5 μg/mL
Formulation PBS
Gene Alias CMD1W, CMH15, Metavinculin, MVCL, vinculin
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 23 amino acids near the center of human Vinculin. < Amino Acid Sequence: VREAFQPQEPDFPPPPPDLEQLR
Purification Method Peptide affinity purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7414
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.