Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241237 0.1 mg
NB435944 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241237 Supplier Novus Biologicals Supplier No. NBP241237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vinculin Polyclonal specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Vinculin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias CMD1W, CMH15, Metavinculin, MVCL, vinculin
Gene Symbols VCL
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human Vinculin. The immunogen is located within amino acids 1060 - 1110 of Vinculin . Amino Acid Squence: LGRTNISDEESEQATE
Molecular Weight of Antigen 120 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7414
Test Specificity Vinculin antibody is human, mouse and rat specific. At least three isoforms of Vinculin are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.