Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB201454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201454 Supplier Novus Biologicals Supplier No. NBP241274FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vinculin Polyclonal antibody specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Vinculin
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CMD1W, CMH15, Metavinculin, MVCL, vinculin
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 23 amino acids near the center of human Vinculin. < Amino Acid Sequence: VREAFQPQEPDFPPPPPDLEQLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7414
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.