Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Antibody (hVIN-1), Novus Biologicals™
SDP

Catalog No. NB6001293 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001293 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001293 Supplier Novus Biologicals Supplier No. NB6001293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

Vinculin Monoclonal specifically detects Vinculin in Human, Mouse, Rat, Amphibian, Bovine, Canine, Chicken, Turkey samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Single Cell Western.

Specifications

Antigen Vinculin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone hVIN-1
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:400, Simple Western, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10 - 1:500, Single Cell Western 1:30
Gene Accession No. P18206
Gene Alias CMD1W, CMH15, Metavinculin, MVCL, vinculin
Gene Symbols VCL
Host Species Mouse
Immunogen Purified human vinculin from uterus.
Molecular Weight of Antigen 116 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7414
Test Specificity Specifically labels vinculin at cell-cell and cell-substrate contacts. Shows cross-reactivity with smooth muscle metavinculin.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Canine, Chicken, Turkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.