Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Antibody, Novus Biologicals™
SDP

Catalog No. NBP220859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220859 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220859 Supplier Novus Biologicals Supplier No. NBP220859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vinculin Polyclonal specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Vinculin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.93mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
Gene Accession No. P18206
Gene Alias CMD1W, CMH15, Metavinculin, MVCL, vinculin
Gene Symbols VCL
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Vinculin. The exact sequence is proprietary.
Molecular Weight of Antigen 124 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7414
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Chicken (93%), Rat (98%), Bovine (98%), Xenopus tropicalis (90%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.