Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylphosphonic acid, 90%, technical, Thermo Scientific Chemicals

Catalog No. p-7077225
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC300590100 10 g
AC300590500 50 g
AC300592500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC300590100 Supplier Thermo Scientific Chemicals Supplier No. 300590100
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1746-03-8
Molecular Formula C2H3O3P
Molecular Weight (g/mol) 106.02
MDL Number MFCD00043866
InChI Key ZTWTYVWXUKTLCP-UHFFFAOYSA-L
Synonym vinylphosphonic acid, phosphonic acid, ethenyl-, homopolymer, vinyl-phosphonicaci, ethenephosphonicacid, ethenylphosphonicacid, ethenyl-phosphonicaci, ethylenephosphonicacid, phosphonic acid, p-ethenyl-, homopolymer, acmc-1bvr6, poly vinylphosphonic acid
PubChem CID 168725
IUPAC Name ethenylphosphonic acid
SMILES [O-]P([O-])(=O)C=C

Specifications

Melting Point 36.0°C
Density 1.3700g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Acid)
Linear Formula H2C=CHP(O)(OH)2
Quantity 10 g
Specific Gravity 1.37
Solubility Information Solubility in water: soluble.
Formula Weight 108.03
Percent Purity 90%
Grade Technical
Chemical Name or Material Vinylphosphonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED
EINECSNumber 217-123-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.