Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyltrimethylsilane, 98+%

Catalog No. p-7046969
Click to view available options
Quantity:
25 g
100 g

CAS: 754-05-2 | C5H12Si | 100.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 754-05-2
Molecular Formula C5H12Si
Molecular Weight (g/mol) 100.236
MDL Number MFCD00008606
InChI Key GCSJLQSCSDMKTP-UHFFFAOYSA-N
Synonym vinyltrimethylsilane, trimethyl vinyl silane, trimethylvinylsilane, silane, ethenyltrimethyl, trimethylsilyl ethylene, vinyl trimethylsilane, silane, trimethylvinyl, vinyltrimethysilane, trimethylsilyl ethene, ethenyl-trimethylsilane
PubChem CID 79102
IUPAC Name ethenyl(trimethyl)silane
SMILES C[Si](C)(C)C=C

Specifications

Boiling Point 54°C to 56°C
Melting Point -132°C
Quantity 25 g
UN Number UN1993
Beilstein 956572
Flash Point −24°C (−11°F)
Formula Weight 100.24
Percent Purity ≥98%
Odor Mild
Refractive Index 1.391
Density 0.69
Chemical Name or Material Vinyltrimethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-042-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.