Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIP Antibody (OTI1E8), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274848J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274848J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274848J646 Supplier Novus Biologicals Supplier No. NBP274848JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VIP Monoclonal specifically detects VIP in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen VIP
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E8
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides
Gene Symbols VIP
Host Species Mouse
Immunogen Full length recombinant protein of human VIP (NP_003372) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7432.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.