Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viperin Antibody, Novus Biologicals™
SDP

Catalog No. NBP203971SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP203971SS 0.025 mg
NBP203971 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP203971SS Supplier Novus Biologicals Supplier No. NBP203971SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Viperin Polyclonal specifically detects Viperin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Chinese Hamster, Equine, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Viperin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_542388.2
Gene Alias 2510004L01Rik, cig33, cig5, Cytomegalovirus-induced gene 5 protein, interferon-inducible, radical S-adenosyl methionine domain containing 2, radical S-adenosyl methionine domain-containing protein 2, vig1, Viperin, Virus inhibitory protein, endoplasmic reticulum-associated
Gene Symbols RSAD2
Host Species Rabbit
Immunogen A portion of amino acids 165-215 of human viperin was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91543
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, white-cheeked gibbon, Chinese hamster, rhesus monkey, common chimpanzee, pig, new world monkey, panda, african bush elephant and orangutan, 95% in horse, wild turkey, opossum, platypus, 89% in western clawed frog, rabbit, zebra finch, chicken, 79% in carolina anole, and 63% homologous in zebrafish and sablefish.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Chinese Hamster, Equine, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.