Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viperin Mouse anti-Human, Mouse, Clone: MaP.VIP, Novus Biologicals™
SDP

Catalog No. NB173638 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173638 100 μg
NB413724 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB173638 Supplier Novus Biologicals Supplier No. NBP307918100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Viperin Monoclonal antibody specifically detects Viperin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence

Specifications

Antigen Viperin
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Clone MaP.VIP
Concentration 0.1 mg/ml
Conjugate Unconjugated
Formulation PBS containing 0.1% BSA
Gene Alias 2510004L01Rik, cig33, cig5, Cytomegalovirus-induced gene 5 protein, interferon-inducible, radical S-adenosyl methionine domain containing 2, radical S-adenosyl methionine domain-containing protein 2, vig1, Viperin, Virus inhibitory protein, endoplasmic reticulum-associated
Host Species Mouse
Immunogen Recombinant mouse viperin fragment (residues 92-362)
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 91543
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.