Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50
Specifications
Antigen | HIV-1 Gag p24 |
---|---|
Clone | 1042960 |
Dilution | Western Blot 1 ug/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11188SP
![]() |
R&D Systems
MAB11188SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11188100
|
R&D Systems
MAB11188100 |
100 μg | N/A | N/A | N/A | ||||
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISASpecifications
HIV-1 Gag p24 | |
Western Blot 1 ug/mL, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
155030 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1042960 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Virus | |
Q77YG1_9HIV1 | |
E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1_9HIV1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title