Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Zika Virus (H/PF/2013) Envelope Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
| Antigen | Zika Virus (H/PF/2013) Envelope |
|---|---|
| Clone | 990844 |
| Dilution | Western Blot 1 μg;/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB100091SP
![]() |
R&D Systems
MAB100091SP |
25 μg |
Each for $200.50
|
|
|||||
MB100091100
![]() |
R&D Systems
MAB100091100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Zika Virus (H/PF/2013) Envelope Monoclonal specifically detects Zika Virus (H/PF/2013) Envelope in Viral samples. It is validated for Western Blot, ELISA.Specifications
| Zika Virus (H/PF/2013) Envelope | |
| Western Blot 1 μg;/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Virus | |
| E. coli-derived recombinant Zika Virus (H/PF/2013) Envelope, Leu590-Ser695 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects Zika Virus (H/PF/2013) Envelope (E3) in direct ELISAs. |
| 990844 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title