Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

visinin-like 1 Antibody (2D11), Novus Biologicals™
SDP

Catalog No. NBP225166SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225166SS 0.025 mL
NBP225166 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225166SS Supplier Novus Biologicals Supplier No. NBP225166SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

visinin-like 1 Monoclonal specifically detects visinin-like 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.

Specifications

Antigen visinin-like 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Classification Monoclonal
Clone 2D11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry Free-Floating
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P62760
Gene Alias HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1
Gene Symbols VSNL1
Host Species Mouse
Immunogen Full length human visinin-like 1 [UniProt# P62760]
Molecular Weight of Antigen 22 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7447
Test Specificity Human, Bovine, Mouse and Rat. Other species have not been tested.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.