Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

visinin-like 1 Antibody (3A9), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP225167UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225167UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225167UV Supplier Novus Biologicals Supplier No. NBP225167UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

visinin-like 1 Monoclonal specifically detects visinin-like 1 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen visinin-like 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3A9
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1
Gene Symbols VSNL1
Host Species Mouse
Immunogen Full length human visinin-like 1 [UniProt# P62760]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7447
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.