Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA/B7-H5/PD-1H Antibody (730801) - Azide and BSA Free, R&D Systems™
SDP

Catalog No. MAB71262SP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB71262SP 25 μg
MAB71262 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB71262SP Supplier Novus Biologicals Supplier No. MAB71262SP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for ICC/IF (Negative), Western Blot.

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications ICC/IF (Negative), Western Blot
Classification Monoclonal
Clone 730801
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ICC/IF (Negative)
Formulation PBS with No Preservative
Gene Alias B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Gene Symbols C10ORF54
Host Species Mouse
Immunogen Partial recombinant human VISTA/B7-H5 (amino acids 33-194) [UniProt Q9H7M9]
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.