Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA/B7-H5/PD-1H Antibody (730804), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346678 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346678 Supplier Novus Biologicals Supplier No. FAB71261MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 730804
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession # Q9H7M9
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.