Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA/B7-H5/PD-1H Antibody (855328), Allophycocyanin, R&D Systems™
SDP

Catalog No. MAB9057APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9057APC 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB9057APC Supplier Novus Biologicals Supplier No. MAB9057APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface).

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface)
Classification Monoclonal
Clone 855328
Conjugate APC
Dilution Flow Cytometry, Flow (Cell Surface)
Formulation PBS with 0.05% Sodium Azide
Gene Alias B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Gene Symbols C10ORF54
Host Species Mouse
Immunogen Partial recombinant human VISTA/B7-H5 protein (aa 33-194)
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.