Learn More
VISTA/B7-H5/PD-1H Antibody (855328) - Azide and BSA Free, R&D Systems™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | VISTA/B7-H5/PD-1H |
| Applications | CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface) |
| Classification | Monoclonal |
| Clone | 855328 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formulation | PBS with No Preservative |
| Gene Alias | B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR |
| Gene Symbols | C10ORF54 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.