Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (855328) - Azide and BSA Free, R&D Systems™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | VISTA/B7-H5/PD-1H |
---|---|
Clone | 855328 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface), CyTOF-ready.Specifications
VISTA/B7-H5/PD-1H | |
1.0 mg/mL | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
C10ORF54 | |
IgG2a | |
Protein A or G purified |
855328 | |
Flow Cytometry, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
64115 | |
Partial recombinant human VISTA/B7-H5 protein (aa 33-194) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title