Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VISTA/B7-H5/PD-1H Antibody (MH5A), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB151958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151958 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151958 Supplier Novus Biologicals Supplier No. NBP311969AF594
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

VISTA/B7-H5/PD-1H Monoclonal antibody specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone MH5A
Conjugate Alexa Fluor 594
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Host Species Armenian Hamster
Immunogen The antibody was generated by immunising Armenian hamsters with mVISTA/B7-H5/PD-1HIg.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 64115
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.