Learn More
VISTA/B7-H5/PD-1H Antibody (MH5A), Janelia Fluor™ 646, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | VISTA/B7-H5/PD-1H |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | MH5A |
| Conjugate | Janelia Fluor 646 |
| Dilution | ELISA, Immunohistochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR |
| Host Species | Armenian Hamster |
| Immunogen | The antibody was generated by immunising Armenian hamsters with mVISTA/B7-H5/PD-1HIg. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.