Learn More
Eisco™ Visual Scientifics Replacement Steel Ball
SCI_ED
Catalog No. p-200007052 Shop All Eisco Products
Click to view available options
Size:
Small
Large
This Visual Scientifics Replacement Steel Ball can be used as a projectile, or in experiments consisting of a free fall or ramp.
  • Replacement steel ball.
Specifications
Product Type Free Fall Apparatus
Material Steel
Size Small
For Use With (Application) Experiments using a projectile, free fall or ramp
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.