Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ VisULite ECL Western Blotting Substrate
Click to view available options
Quantity:
200 mL
Description
- Low picogram level detection
- High signal intensity, duration
- Compatible with X-ray film or CCD imaging systems
Storage: Store in the dark at 2°C to 8°C. Do not freeze.
Specifications
Specifications
Product Type | ECL Western Blot Substrate |
Quantity | 200 mL |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction